Ferrule Gripper Sets, Ace Glass Incorporated
# de catalogue 76318-538
Fournisseur: Ace Glass
Spécifications
- Description:Ferrule/Gripper Set
- Size:1/4"
- Cat. no.:76318-538
Spécifications
A propos de cet article
Replacement ferrule/gripper sets for ACE PTFE tube compression fittings.
Delivery: Each set consists of one each PTFE ferrule and PVDF gripper.
Customers Who Bought This Also Bought