Spécifications
- Description:BD Vacutainer® Luer-Lok™ access device - holder with preattached multiple sample adapter
- Cat. no.:VT36490200
Spécifications
A propos de cet article
The BD Vacutainer® Luer-Lok™ access device provides sterile, secure and safe specimen sampling for fast, accurate and consistent results that can lead to improved patient outcomes.
- Sterile
- Latex-free
- Red colored tip for differentiation
It offers the security of a threaded, Luer locking connection. Compatible with needleless IV sites designed for Luer-lock access, and Luer locking Foley Catheter sampling ports.