ContactUsLinkComponent
Luer-Lok Access Device Vacutainer Cs200
BD Vacutainer® Luer-Lok™ Access Device, BD
# de catalogue VT36490200
Fournisseur:  BD
Luer-Lok Access Device Vacutainer Cs200
BD Vacutainer® Luer-Lok™ Access Device, BD
# de catalogue VT36490200
Fournisseur:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    BD Vacutainer® Luer-Lok™ access device - holder with preattached multiple sample adapter
  • Cat. no.:
    VT36490200
Spécifications
A propos de cet article
The BD Vacutainer® Luer-Lok™ access device provides sterile, secure and safe specimen sampling for fast, accurate and consistent results that can lead to improved patient outcomes.
  • Sterile
  • Latex-free
  • Red colored tip for differentiation
It offers the security of a threaded, Luer locking connection. Compatible with needleless IV sites designed for Luer-lock access, and Luer locking Foley Catheter sampling ports.
Customers Who Bought This Also Bought