Cuvette Washer, FireflySci
# de catalogue 76314-064
Fournisseur: FIREFLYSCI, INC.
Spécifications
- Description:Single Cuvette Washer Kit with Vacuum Pump
- Cat. no.:76314-064
Spécifications
A propos de cet article
Cuvette washers are handmade and built to last for multiple uses without contamination.
- Fast cuvette cleaning action
- Leave cuvettes in place for quick drying action
- Use with cuvettes from 3 to10mm lightpath
- Reduces handling and risk of breakage
Easy to use device requires the cuvette to be turned upside down when the cleaning solution is run.
Ordering information: Washers includes Gasket and stopper. Vacuum flask and pump must be ordered separately. Double cuvette washer kit includes cuvette washer head, rubber gasket, stopper (size 7), filtering flask, and vinyl tubing (304.8 cm). Kits are also available with vacuum pump.
Customers Who Bought This Also Bought