Corning® Media Bottles, PET, with IATA-Validated Screw Cap, Corning
# de catalogue 76312-394
Fournisseur: Corning
Spécifications
- Cap Size:34.7 mm
- Capacity:250 ml
- Sterility:Sterile
- Material:PET bottle/HDPE cap
- Cat. no.:76312-394
- Packaging:Shrink-wrapped tray in outer bag
- Cap Color:Natural
Spécifications
A propos de cet article
Corning® PET media bottles are equipped with a screw cap designed to ensure leak-proof performance.
- Available in four sizes
- Packaged in convenient, shrink-wrapped trays within an outer bag to ensure cleanliness
- Break-resistant bottles are an ergonomic and easy-to-handle alternative to glass
- Molded graduations for accurate measurements
- Innovative HDPE screw cap meets IATA requirements to ensure leak-proof air transport conditions
These bottles with IATA-validated screw caps are designed for safe, secure storage of tissue culture media, sera, buffers, and other aqueous solutions. The PET resin makes the bottle lightweight, break-resistant, and as transparent as glass. The screw cap is designed to ensure leak-proof performance and meets International Air Transport Association (IATA) requirements for primary packaging used in air transportation.
Certifications: Bottles have Sterility Assurance Level (SAL) of 10⁻⁶. Bottles are validated USP Class VI, noncytotoxic, nonhemolytic, and nonpyrogenic.
Customers Who Bought This Also Bought