Hydranal® Coulometric Karl Fischer Titration Reagents, Honeywell Research Chemicals
# de catalogue BJ34868-500MLS
Fournisseur: Honeywell Research Chemicals
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Polycoated Bottle
- Description:HYDRANAL™-Coulomat Oil, Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm
- Size:1 x 500 mL
- Cat. no.:BJ34868-500MLS
Spécifications
A propos de cet article
Coulometric Karl Fischer titrations normally require two reagent solutions: an anolyte (the solution in the anodic compartment) and a catholyte (the solution in the cathodic compartment).
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range available
- Long shelf life (up to five years)
Customers Who Bought This Also Bought