Ward's® White's Tree Frog
# de catalogue 470312-390
Fournisseur: Avantor
Spécifications
- Description:Whites Tree Frog
- Cat. no.:470312-390
- Packaging:Pack of 3
Spécifications
A propos de cet article
Your frog is shipped in a plastic container with damp sphagnum moss as cushioning. It should be placed in a prepared habitat shortly after arrival.
- Litoria caerulea
Ordering information: These products contain living or perishable materials. Visit https://wardsci.com/cms/teacher_live_materials for care instructions.
Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought