ContactUsLinkComponent
Next generation universal wwPTFE membrane disc filter, 47mm 0.2um
wwPTFE (Water Wettable PTFE) Membrane Disc Filters, Cytiva (Formerly Pall Lab)
# de catalogue 76308-706
Fournisseur:  Cytiva
Next generation universal wwPTFE membrane disc filter, 47mm 0.2um
wwPTFE (Water Wettable PTFE) Membrane Disc Filters, Cytiva (Formerly Pall Lab)
# de catalogue 76308-706
Fournisseur:  Cytiva
Spécifications
  • Ø:
    47 mm
  • Pore size:
    0.20 µm
  • Cat. no.:
    76308-706
Spécifications
A propos de cet article
Next generation of universal hydrophilic PTFE membrane with maximum chemical compatibility when filtering both aqueous solutions and aggressive solvents.
  • Hydrophilic
  • Low protein binding
HPLC certified for low extractables assuring that the filter will not add artifacts or contamination to your analysis. Low protein binding of wwPTFE membrane provides high recovery of critical proteinaceous samples.
Customers Who Bought This Also Bought