Ward's Adapt-A-Rack
# de catalogue 470311-050
Fournisseur: VWR International
Spécifications
- Dimensions:18.1×5.6×7.6 cm
- Color:Pink/White and White/Pink
- Cat. no.:470311-050
Spécifications
A propos de cet article
Flexible multi-tube rack.
- Holds different sized tubes in each slot
- Racks can be linked together for additional holding capacity
- Open sides allow for label viewing
- Flat base to hold tubes upright
- Linked racks should be supported underneath when carrying
- Available in color combinations for easy coding and identification
- Autoclavable
Each slot on the Ward's Adapt-A-Rack adapts to accommodate 12 to 30 mm (5 to 50 ml) tube sizes and holds them in a firm upright position. Each rack is made from POM and contains four wells.
Customers Who Bought This Also Bought