Single Scale Graduated Cylinders
# de catalogue 470310-996
Fournisseur: Wards
Spécifications
- Capacity:50 ml
- Graduation Interval:0.5 ml
- Cat. no.:470310-996
Spécifications
A propos de cet article
Single Scale Graduated Cylinders with plastic base and protection collar.
- Class B
- Borosilicate glass gradauted cylinder
- Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
Customers Who Bought This Also Bought