Spécifications
- Description:Vesphene III SE Phenolic Disinfectant
- Size:4×1 gal.
Spécifications
A propos de cet article
Vesphene III se disinfectant phenolic disinfectant is a one-step, alkaline phenolic cleaner and detergent which is specifically formulated for use on hard, non-porous surfaces.
- Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy
- Productivity - Formulation includes surfactants for surface cleaning while disinfecting
- Flexibility - Ideal for use in a contamination control rotation
- Reliability - Complete lot traceability
- Assurance - Each lot documented by certificate of analysis
Vesphene® III st have dilution rate of 0.8% v/v (1:128), kills a broad spectrum of common bacteria and fungi.
The use-dilution can be applied manually or sprayed.