ContactUsLinkComponent
Assembly Filtration
Filtration Assembly
# de catalogue 470310-690
Fournisseur:  Wards
Assembly Filtration
Filtration Assembly
# de catalogue 470310-690
Fournisseur:  Wards
Spécifications
  • Description:
    Filtration Assembly
  • Cat. no.:
    470310-690
Spécifications
A propos de cet article
This filtration assembly is used for vacuum filtration of tissue culture media, biological fluids, HPLC and other aqueous solutions.
  • Unit is designed to maximize flow and reduce foaming
  • Versatile and effective
Assembly consists of 300 ml borosilicate glass funnel, highly integrated sealed 47 mm fritted glass support base, one litre flask. An aluminum clamp is provided to hold the filter funnel and support base properly. The 47 mm filter provides an effective filtration area of 9.6 square centimeters.
Customers Who Bought This Also Bought