KleenGuard™ Maverick™ Safety Glasses, Kimberly-Clark Professional®
# de catalogue 76300-904
Fournisseur: Kimberly-Clark
Spécifications
- Lens Coating:Anti-fog
- Lens Tint:Clear
- Description:Maverick™ safety glasses
- Frame Color:Black
- Cat. no.:76300-904
Spécifications
A propos de cet article
Maverick™ eye protection combines a modern, sleek, and trendy design with reliable comfort and safety features.
- Modern, sleek, stylish, and lightweight design employees are happy to wear
- Integrated browguard and side shields help the user feel confident they are protected
- Comfort touch points on nose bridge and temples prevent slippage and reduce pressure
- Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
- Premium Anti-Fog and Premium Anti-Scratch lens coatings for better performance and longer life
- Eligible for recycling via RightCycle by Kimberly-Clark Professional®
Workers need protection, but they also care how they look. Maverick™ Eye Protection, the first eyewear created for the KleenGuard™ Brand, delivers confident protection without compromising style or comfort. It’s PPE that workers will want to wear.
Certifications: Meets the ANSI Z87.1+ standards for optical and impact protector requirements. Certified to meet the impact, ignition/flammability, coverage, and optical requirements of the CSA Z94.3 standards.
Customers Who Bought This Also Bought