ContactUsLinkComponent
Safety Glasses Clr Lens-Black Frame CS12
KleenGuard™ Maverick™ Safety Glasses, Kimberly-Clark Professional®
# de catalogue 76300-904
Fournisseur:  Kimberly-Clark
Safety Glasses Clr Lens-Black Frame CS12
KleenGuard™ Maverick™ Safety Glasses, Kimberly-Clark Professional®
# de catalogue 76300-904
Fournisseur:  Kimberly-Clark
Spécifications
  • Lens Coating:
    Anti-fog
  • Lens Tint:
    Clear
  • Description:
    Maverick™ safety glasses
  • Frame Color:
    Black
  • Cat. no.:
    76300-904
Spécifications
A propos de cet article
Maverick™ eye protection combines a modern, sleek, and trendy design with reliable comfort and safety features.
  • Modern, sleek, stylish, and lightweight design employees are happy to wear
  • Integrated browguard and side shields help the user feel confident they are protected
  • Comfort touch points on nose bridge and temples prevent slippage and reduce pressure
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
  • Premium Anti-Fog and Premium Anti-Scratch lens coatings for better performance and longer life
  • Eligible for recycling via RightCycle by Kimberly-Clark Professional®
Workers need protection, but they also care how they look. Maverick™ Eye Protection, the first eyewear created for the KleenGuard™ Brand, delivers confident protection without compromising style or comfort. It’s PPE that workers will want to wear.
Certifications: Meets the ANSI Z87.1+ standards for optical and impact protector requirements. Certified to meet the impact, ignition/flammability, coverage, and optical requirements of the CSA Z94.3 standards.
Customers Who Bought This Also Bought