KleenGuard™ A45 Liquid and Particle Protection Surface Prep and Paint Apparel, Kimberly-Clark Professional
# de catalogue 76300-898
Fournisseur: Kimberly-Clark
Spécifications
- Anti-static:Yes
- Clothing Size:3XL
- Garment application:Industrial
- Color:White
- Description:Zipper front, elastic wrists, ankles, hood and boots
- Model:A45
- Cat. no.:76300-898
Spécifications
A propos de cet article
The KleenGuard™ A45 liquid and particle protection coveralls for surface prep and paint are designed to protect workers where hazardous or non-hazardous contaminants may be present.
- Multiple styles are available
- Made of microporous film laminate with a water-resistant zipper front
- REFLEX coverall design offers excellent fit and freedom of movement, so workers won't need to modify them to do their job
- Coveralls with a hood are designed to fit a respirator
- Protects against chromate dust (10%) and isocyanates (40% concentration), hazardous materials often used in industrial paint jobs
- Skid-resistant boots provide up to 68% improvement in traction on wet and dry surfaces when compared to former boot materials
The KleenGuard™ A45 liquid and particle protection coveralls for surface prep and paint are designed to move with workers where hazardous or non-hazardous contaminants may be present. Each protective suit is made with lightweight fabric, allowing for improved comfort without sacrificing protection. They are not made with natural rubber latex and are silicone-free, and they feature serged seams and a water resistant zipper.
Certifications: Passes NFPA 99 criteria for antistatic materials.
Customers Who Bought This Also Bought