VWR® Sterile T-Shaped Cell Spreaders
Catalog # 76299-568
Supplier: VWR International
Specifications
- Sterility:Sterile
- Color:Purple
- Description:T-Shape Spreader
- Cat. no.:76299-568
- Packaging:1/Peel Bag
Specifications
About this item
VWR® disposable T-shaped Cell Spreaders are ideal for spreading bacterial cultures evenly.
- Spread and disperse liquids across the surface of culture plates
- Smooth edges minimize 'gouging'
- Sterilized by EO irradiation
- BSE/TSE-free, and latex-free
VWR® T-Shaped Cell Spreaders are manufactured under EN ISO 13485:2016 and ISO 9001:2015 quality management systems.
Certifications: ISO 13485 :2003 and ISO 9001: 2008.
Customers Who Bought This Also Bought