ContactUsLinkComponent
VWR cell spreader, T-shaped, Purple, 10/peel bag, 100bags/Box, 6 Boxes/Case, EO
VWR® Sterile T-Shaped Cell Spreaders
# de catalogue 76299-566
Fournisseur:  VWR International
VWR cell spreader, T-shaped, Purple, 10/peel bag, 100bags/Box, 6 Boxes/Case, EO
VWR® Sterile T-Shaped Cell Spreaders
# de catalogue 76299-566
Fournisseur:  VWR International
Spécifications
  • Sterility:
    Sterile
  • Color:
    Purple
  • Description:
    T-Shape Spreader
  • Cat. no.:
    76299-566
  • Packaging:
    10/Peel Bag
Spécifications
A propos de cet article
VWR® disposable T-shaped Cell Spreaders are ideal for spreading bacterial cultures evenly.
  • Spread and disperse liquids across the surface of culture plates
  • Smooth edges minimize 'gouging'
  • Sterilized by EO irradiation
  • BSE/TSE-free, and latex-free
VWR® T-Shaped Cell Spreaders are manufactured under EN ISO 13485:2016 and ISO 9001:2015 quality management systems.
Certifications: ISO 13485 :2003 and ISO 9001: 2008.
Customers Who Bought This Also Bought