Neoprene Stoppers, United Scientific Supplies
# de catalogue 76293-792
Fournisseur: United Scientific Supplies
Spécifications
- Size:8
- Cat. no.:76293-792
- Top Diameter:41 mm
- Bottom Diameter:33 mm
- No./lb.:12
Spécifications
A propos de cet article
Synthetic, neoprene stoppers contain no free sulfur and exhibit good resiliency and shelf-life.
- Neoprene fabrication
Neoprene stoppers can be used in areas where rubber would be attacked by reagents or solvents. Synthetic green neoprene stoppers have good resiliency and shelf-life. They withstand petroleum products and oils as well as most inorganic acids and bases.
Packaging: Sold as one-pound bags.
Customers Who Bought This Also Bought