Steel Rod Support Rings, United Scientific Supplies
Catalog # 76293-654
Supplier: United Scientific Supplies
Specifications
- Ext.Ø:4"
- Cat. no.:76293-654
Specifications
About this item
Steel rings for holding funnels.
- Steel rod, zinc plated
- Boss head clamp
Steel rod, zinc-plated support ring with attached boss head clamp and thumbscrew. Distance from support to center of ring is 4-1/2" for all sizes.
Customers Who Bought This Also Bought