Order Entry
ContactUsLinkComponent
Support Stand Rod Stamped Steel 4Inx6In
Stamped Steel Support Stands with Rods, United Scientific Supplies
# de catalogue 76293-616
Support Stand Rod Stamped Steel 4Inx6In
Stamped Steel Support Stands with Rods, United Scientific Supplies
# de catalogue 76293-616

Spécifications

  • Cat. no.:
    76293-616
  • Base Dimensions:
    4×6"
  • Rod Diameter:
    ⁵⁄₁₆"
  • Rod Height:
    18"

Spécifications

A propos de cet article

Stamped steel bases, with black enamel finish.

  • Zinc plated steel rod

Sturdy steel base.

Ordering information: Bases and Rods are also available individually.
Delivery: Includes a removable, zinc-plated steel rod.
Customers Who Bought This Also Bought