Watch Glasses, United Scientific Supplies
# de catalogue 76293-562
Fournisseur: United Scientific Supplies
Spécifications
- Ø:100 mm
- Description:Watch glass
- Cat. no.:76293-562
Spécifications
A propos de cet article
Designed to perform evaporations on a hot plate or over open flame.
- Soda lime glass
- Flame polished edges
Made from soda lime glass (1.8 mm thick), with fine ground and flame polished edges.
Customers Who Bought This Also Bought