ContactUsLinkComponent
Crucible Porous Bottom Porcelain 5.0 Um
Porcelain Crucibles with Porous Bottom, United Scientific Supplies
# de catalogue 76293-546
Crucible Porous Bottom Porcelain 5.0 Um
Porcelain Crucibles with Porous Bottom, United Scientific Supplies
# de catalogue 76293-546
Spécifications
  • Capacity:
    25 ml
  • Cat. no.:
    76293-546
  • Pore Diameter:
    5 µm
Spécifications
A propos de cet article
Porous bottom crucibles have a molded-in porous disc with stable porosity and consistent flow rate.
  • Porous bottom
  • Glazed
Available with very fine (1.2 µm), fine (5 µm), and medium (15 µm) pore diameters. Glazed inside and out, except for the outside bottom and rim. Autoclavable.
Customers Who Bought This Also Bought