Graham Condensers, 24/40 Joint, United Scientific Supplies
# de catalogue 76293-452
Fournisseur: United Scientific Supplies
Spécifications
- Length:300 mm
- ST Joint:24/40
- Cat. no.:76293-452
Spécifications
A propos de cet article
Graham condensers are designed for use in distillation applications.
- 24/40 sockets
- Screw thread connector
Constructed of borosilicate glass, this condenser has a coiled inner tube to provide additional surface area for highly efficient cooling. Socket is 24/40 with a screw thread connector.
Customers Who Bought This Also Bought