Test Tube Clamps, United Scientific Supplies
# de catalogue 76293-444
Fournisseur: United Scientific Supplies
Spécifications
- Description:Steel Wire, Brass Finish, without Finger Grips
- Cat. no.:76293-444
Spécifications
A propos de cet article
Pick up test tubes easily and safely.
- Molded finger grips for easy manipulation
- Heavy gauge wire with one-piece construction
- Length: 5"
Self-closing, available in brass wire or steel wire with brass finish.
Customers Who Bought This Also Bought