ContactUsLinkComponent
Test Tube Clamp, Steel Wire, Brass Finish, Without Finger Grips, Self-closing
Test Tube Clamps, United Scientific Supplies
# de catalogue 76293-444
Test Tube Clamp, Steel Wire, Brass Finish, Without Finger Grips, Self-closing
Test Tube Clamps, United Scientific Supplies
# de catalogue 76293-444
Spécifications
  • Description:
    Steel Wire, Brass Finish, without Finger Grips
  • Cat. no.:
    76293-444
Spécifications
A propos de cet article
Pick up test tubes easily and safely.
  • Molded finger grips for easy manipulation
  • Heavy gauge wire with one-piece construction
  • Length: 5"
Self-closing, available in brass wire or steel wire with brass finish.
Customers Who Bought This Also Bought