Deluxe Burette Clamps, United Scientific Supplies
# de catalogue 76293-394
Fournisseur: United Scientific Supplies
Spécifications
- Description:Double Deluxe Burette Clamps
- Cat. no.:76293-394
Spécifications
A propos de cet article
Deluxe Burette clamps are die casted from aluminum and feature black rubber sleeves for firm gripping of burettes.
- Rubber sleeves
- Holds burettes up to 30 mm O.D.
The adjustable clamps hold up to 30 mm O.D. burettes, which are held in place by the pressure created by the springs. For use with rods up to 19 mm diameter.
Customers Who Bought This Also Bought