Order Entry
ContactUsLinkComponent
Support Ring Extension W/Steel Rod 4 In
Extension Support Rings, United Scientific Supplies
# de catalogue 76293-338
Support Ring Extension W/Steel Rod 4 In
Extension Support Rings, United Scientific Supplies
# de catalogue 76293-338

Spécifications

  • O.D.:
    4"
  • Cat. no.:
    76293-338

Spécifications

A propos de cet article

Extension support for holding funnels.

  • Steel rod, zinc plated
  • 12" long

Steel rod, zinc-plated extension support rings. Rod diameter 5/16", overall length 12" for all sizes.

Customers Who Bought This Also Bought