Stainless Steel Laboratory Scoop, United Scientific Supplies
Catalog # 76293-326
Supplier: United Scientific Supplies
Specifications
- Description:Laboratory Scoop, Stainless Steel
- Cat. no.:76293-326
Specifications
About this item
Great quality stainless steel scoop with one pointed end for handling small amounts of chemicals.
- Length is 6.5"
- Also available with a 5" long hardwood handle
The stainless steel scoop has a narrow design for inserting into bottles. Perfect for removing small amounts of powdered material.
Useful for portioning out powders from small containers.
Customers Who Bought This Also Bought