Spécifications
- Capacity:12 ml
- Description:Exel Curved Tip Syringe
- Cat. no.:76290-492
- Packaging:50/box
- Type:Curved dispensing tip
Spécifications
A propos de cet article
EXEL international's quality disposable curved tip dispensing syringe. Great value at reasonable costs.
- Standard three part syringe
- Latex and pyrogen free
- Non-toxic
- Positive plunger stop
EXEL international is known for unsurpassed quality at a low price. The design includes bold print, clear barrels, and siliconized gaskets for smooth, consistent plunger motion and a positive plunger stop.