Spécifications
- Length:1/2"
- Volume:³⁄₁₀ ml
- Description:Syringes with Fixed Needles
- Sterility:Sterile
- Packaging:100/box
- For Use With:Human, Laboratory, and Veterinary Use
- Gauge:29
- Type:Insulin
Spécifications
A propos de cet article
These EXEL International Insulin Syringes with Fixed Needle are high quality and cost-efficient.
- Sterile
- Latex free
- Non pyrogenic
- Non toxic
- Low deadspace
The insulin syringes design includes bold printing, clear graduations and siliconized gaskets for smooth consistent plunger motion and a positive plunger stop. The auto ground needle tip provides for greater comfort. Reduced dead space in this design makes for reduced Insulin waste.