Spécifications
- Capacity:5 ml
- Sterility:Sterile
- Description:Exel Disposable Syringes
- Cat. no.:76290-380
- Packaging:100/box
- For Use With:Human, Laboratory, and Veterinary use
- Type:Luer Lock
Spécifications
A propos de cet article
EXEL international's quality disposable syringes. Great value at reasonable costs.
- Standard three part syringe
- Latex and pyrogen free
- Non-toxic
- Positive plunger stop
EXEL international is known for unsurpassed quality at a low price. The design includes bold print, clear barrels, and siliconized gaskets for smooth, consistent plunger motion and a positive plunger stop.