Spécifications
- Capacity:10 ml
- Description:Exel Disposable Syringes
- Cat. no.:76290-374
- Packaging:100/box
- For Use With:Human, Laboratory, and Veterinary use
- Type:Luer Slip
Spécifications
A propos de cet article
EXEL international's quality disposable syringes. Great value at reasonable costs.
- Standard three part syringe
- Latex and pyrogen free
- Non-toxic
- Positive plunger stop
EXEL international is known for unsurpassed quality at a low price. These syringes are latex free, sterile, non-pyrogenic and non-toxic. The design includes bold print, clear barrels, and siliconized gaskets for smooth, consistent plunger motion and a positive plunger stop.