ContactUsLinkComponent
Test Strip Dual Range Qac Treated 50Test
Dual Range QAC Test Strips, LaMotte®
# de catalogue 470308-466
Fournisseur:  LaMotte
Numéro CAS:  
Test Strip Dual Range Qac Treated 50Test
Dual Range QAC Test Strips, LaMotte®
# de catalogue 470308-466
Fournisseur:  LaMotte
Numéro CAS:  
Spécifications
  • Packaging:
    50/vial, 1 vial
  • Description:
    Dual Range QAC Test Strips
  • Cat. no.:
    470308-466
  • Range:
    0 - 800
Spécifications
A propos de cet article
The New Dual Range QAC Test Strip takes care of ALL of your testing needs on ONE STRIP!
  • Low Range: 0, 10, 20, 40, 80
  • High Range: 0, 100, 200, 400, 800
Whether checking the concentration of your QAC solution or checking to ensure complete removal, the new Dual Range QAC Test Strip DOES IT ALL!
Chemically-treated test strips change color to indicate sanitizer level. Strips and color chart are packaged in a waterproof plastic vial.
Customers Who Bought This Also Bought