Dual Range QAC Test Strips, LaMotte®
# de catalogue 470308-466
Fournisseur: LaMotte
Numéro CAS:
Spécifications
- Packaging:50/vial, 1 vial
- Description:Dual Range QAC Test Strips
- Cat. no.:470308-466
- Range:0 - 800
Spécifications
A propos de cet article
The New Dual Range QAC Test Strip takes care of ALL of your testing needs on ONE STRIP!
- Low Range: 0, 10, 20, 40, 80
- High Range: 0, 100, 200, 400, 800
Whether checking the concentration of your QAC solution or checking to ensure complete removal, the new Dual Range QAC Test Strip DOES IT ALL!
Chemically-treated test strips change color to indicate sanitizer level. Strips and color chart are packaged in a waterproof plastic vial.
Customers Who Bought This Also Bought