ContactUsLinkComponent
Eye Pad W/Adhesive Strips First Aid 4/BX
First Aid Only™ Eye Pads and Strips, Acme United
# de catalogue 76287-282
Fournisseur:  First Aid Only
Eye Pad W/Adhesive Strips First Aid 4/BX
First Aid Only™ Eye Pads and Strips, Acme United
# de catalogue 76287-282
Fournisseur:  First Aid Only
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Eye Pads and Strips
  • Cat. no.:
    76287-282
Spécifications
A propos de cet article
Sterile Eye Pads for eye injuries.
  • 8-piece eye pad and adhesive strip kit
Individually packaged eye pads and adhesive strips are designed for use with eye injuries. Sold as box of four pads and four strips. Perfect for stand-alone first aid or as a refill for a kit.
Customers Who Bought This Also Bought