MycoSensor qPCR Assay Kits, Agilent Technologies
MycoSensor qPCR Assay Kits, Agilent Technologies
MycoSensor qPCR assay kit, All the reagents you need for amplification and fluorescence detection, internal controls, DNA purification reagents., 50 reactions
Catalog # CAST302107
Supplier #:  302107
MycoSensor qPCR Assay Kits, Agilent Technologies
Catalog # CAST302107
CAS Number:  
MycoSensor qPCR Assay Kits, Agilent Technologies
MycoSensor qPCR Assay Kits, Agilent Technologies
MycoSensor qPCR assay kit, All the reagents you need for amplification and fluorescence detection, internal controls, DNA purification reagents., 50 reactions
CAS Number:  
Price
$396.66
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The MycoSensor PCR and qPCR assay kits detect cultured Mycoplasma infection by real-time qPCR or PCR.
  • Detects Mycoplasma infection of cell cultures by qPCR or PCR in less than two hours
  • Specific detection of the eight most common Mycoplasma species
  • SYBR Green dye detection
The MycoSensor PCR and qPCR assay kits detect Mycoplasma infection of cell cultures by real-time quantitative PCR (qPCR) or PCR in less than two hours. The easy-to-use system can specifically detect the eight most common Mycoplasma species.
The system uses SYBR Green dye detection. The kit includes a primer mix that amplifies the eight most common Mycoplasma species that infect cell cultures. The MycoSensor qPCR assay kit primer mix can detect Mycoplasma infections in cell cultures in less than two hours.
Specifications
Specifications
  • Désignation:
    MycoSensor qPCR assay kit
  • Taille:
    50 reactions
  • Details:
    All the reagents you need for amplification and fluorescence detection, internal controls, DNA purification reagents.