ContactUsLinkComponent
WARDS WEIGHING PAPERS 4X4IN WH PK/500
Ward's® Nitrogen-Free Weighing Papers
Catalog # 470307-002
WARDS WEIGHING PAPERS 4X4IN WH PK/500
Ward's® Nitrogen-Free Weighing Papers
Catalog # 470307-002
Specifications
  • Size:
    4×4"
  • Cat. no.:
    470307-002
  • Packaging:
    500 sheets/pack
Specifications
About this item
Nitrogen-free papers feature a very smooth surface for excellent sample recovery.
They are strong enough to support even dense materials.
Packaging: Pack of 500.
Customers Who Bought This Also Bought