Order Entry
ContactUsLinkComponent
Marker Cryo Alcohol-Water-Resistant PK1
SCIENCE-MARKER™ Cryogenic Markers, GA International
Catalog # 76276-060
undefined
SCIENCE-MARKER™ Cryogenic Markers, GA International
Catalog # 76276-060

Specifications

  • Color:
    Black
  • Description:
    Cryogenic permanent marker
  • Dimensions:
    15.9×101.6×168.3 mm
  • Cat. no.:
    76276-060

Specifications

About this item

Cryogenic permanent marker with fine tip.

  • Alcohol and water resistant
  • Compatible with Lab-TAG™ labels

Shipping weight 113 g

Delivery: Supplied six markers per pack.
Caution: Not suitable for exposure to Acid/Ethanol mixes.
Frequently Bought Together