Order Entry
ContactUsLinkComponent
Cryogenic Label White 0.43in 1 pk
Permanent Cryogenic Labels for Laser Printers, US Letter Size
# de catalogue 76271-670
Fournisseur:  GA INTERNATIONAL INC
Cryogenic Label White 0.43in 1 pk
Permanent Cryogenic Labels for Laser Printers, US Letter Size
# de catalogue 76271-670
Fournisseur:  GA INTERNATIONAL INC

Spécifications

  • Color:
    White
  • Description:
    US letter size - Cryogenic labels for laser printers
  • Packaging:
    5376 labels/pack (336 labels/sheet)
  • Dimensions:
    11 mm (0.43")

Spécifications

A propos de cet article

Permanent cryogenic laser labels for long-term storage in liquid-phase and vapor phase liquid nitrogen (–196 °C), freezers (–80 °C), and on dry ice. Supplied in sheet format for printing in standard desktop laser printers.

  • Temperature range: –196 to +120 °C (–321 to +248 °F)
  • Strong permanent adhesive ensures labels will not wrinkle or peel
  • Resists extreme cryogenic temperatures (–196 °C in a liquid phase and vapor phase liquid mnitrogen)
  • Printable by all major desktop laser printers
  • Resistant to gamma irradiation (tested to up to 50 kGy)

Coated with a unique cryo adhesive, thermoplastic waterproof labels are specifically designed to permanently adhere to vials, microtubes, microplates, glass tubes, freezer boxes, polycarbonate cryo boxes, and other containers that will undergo long-term deep-freeze and cryogenic storage.

Provided in sheet format for printing with desktop laser printers, these cryogenic tags are wear- and tear-proof. They will not jam printers, and are resistant to UV and Gamma irradiation.

Customers Who Bought This Also Bought