Order Entry
ContactUsLinkComponent
Cryolabel White 0.35In Circle Pack of 1
Permanent Cryo-LazrTAG™, Deep-Freeze Labels for Laser Printers, GA International
Catalog # 76271-620
undefined
Permanent Cryo-LazrTAG™, Deep-Freeze Labels for Laser Printers, GA International
Catalog # 76271-620

Specifications

  • Color:
    White
  • Description:
    Round cryo labels on sheets for laser printing
  • Dimensions:
    0.35"
  • Cat. no.:
    76271-620
  • Core Size:
    1"

Specifications

About this item

Deep-freeze thermoplastic laser labels for long-term storage in –80 °C freezers.

  • Available in a wide range of colors; perfect for color-coding
  • Labels withstand sterilisation by steam, dry heat, and gamma irradiation
  • Strong permanent adhesive ensures labels won't wrinkle or peel
  • Printable by all major desktop laser printers
  • Resistant to gamma irradiation (tested to up to 50 kGy)

These circular labels are provided in a range of colors and accept laser printing and writing with permanent cryo markers. Designed for the identification of 0.5, 1.5, and 2 ml microtube and vial caps. Water-resistant, wear and tear proof, these labels will not jam printers. They are resistant to autoclaving, UV, and Gamma irradiation.

Frequently Bought Together