Order Entry
ContactUsLinkComponent
SureSlide* Standard Microscope Slide, 45 Deg Clipped Corner, Colour: White, Dimension: 75 x 25 mm
SureSlide™ Clipped Corner Microscope Slides, Avantik
Catalog # 76270-504
Supplier:  Avantik
SureSlide* Standard Microscope Slide, 45 Deg Clipped Corner, Colour: White, Dimension: 75 x 25 mm
SureSlide™ Clipped Corner Microscope Slides, Avantik
Catalog # 76270-504
Supplier:  Avantik

Specifications

  • Color:
    White
  • Description:
    Slides, 45° clipped corners, colored ends
  • L×W:
    75×25 mm
  • Cat. no.:
    76270-504
  • Packaging:
    72 Slides/Pack

Specifications

About this item

SureSlide™ microscope slides provide consistent and reliable results.

  • Optimized to ensure consistent barcode scanning
  • 100% quality guarantee

These slides are compatible with all major manufacturer automated printers; resulting in superior print quality.

Customers Who Bought This Also Bought