Convenience Kit SureStop Screw Vial 2mL
# de catalogue MP75852-612
Fournisseur: Thermo Scientific
Customers Who Bought This Also Bought
Cerium(IV) sulfate solution 0.1 N, VWR Chemicals BDH®
Fournisseur: VWR International
$470.52 to $5,298.64
Phenolphthalein 1% (w/v) in 95% (v/v) alcohol indicator, neutralized
Fournisseur: Ricca Chemical
$93.96
Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
$330.55 to $1,952.15