ContactUsLinkComponent
XTIP4* Filter Pipette Tip 1000ul
xTIP4™ Racked Pipette Tips, Rainin® LTS® Pipette Compatible, Biotix
Catalog # 76266-146
Supplier:  Biotix
XTIP4* Filter Pipette Tip 1000ul
xTIP4™ Racked Pipette Tips, Rainin® LTS® Pipette Compatible, Biotix
Catalog # 76266-146
Supplier:  Biotix
Specifications
  • Volume:
    1000 µl
  • Packed:
    Racked: 768/Pack, 3840/Case
  • Cat. no.:
    76266-146
  • Sterile:
    +
Specifications
About this item
Biotix xTIP4 pipette tips are fully compatible with Rainin LTS single, multi, and electronic pipettes. Create a perfect ergonomic system in your lab with FlexFit®, StarStop®, and a Biotix xPIPETTE or Rainin LTS® pipette.
  • Polyethylene filters protect against aerosol and liquid contamination with no colorant or CMC contamination
  • Full multi-channel and single-channel LTS and xPIPETTE compatibility
Light Insertion Force
FlexFit® sealing technology requires minimal insertion force to generate consistent seals that users can trust with every tip.
Light Ejection
No matter how tips are applied, xTIP4™ pipette tips will always eject at the same light force. The StarStop® feature works with LTS-style pipettes to create a strong seal in the same location, every time.
Low Binding
Biotix low retention technology reduces the amount of sample lost in the tip when pipetting. Great for use with costly reagenst like proteins, and DNA to increase an experiment's accuracy and yield.
Clean Delivery
These tips are engineered with a Blade® tip to reduce hanging droplet formation when dispensing a sample. This limits the need for tip touch off on plate wells or tubes, saving time and sample.
Customers Who Bought This Also Bought