Empty Optimus Racks for CleanPak™ and TipEject™ Pipette Tip Reloads, Biotix
# de catalogue 76266-138
Fournisseur: Biotix
Spécifications
- Description:Empty Optimus Rack for 10-300 µl Tips
- Cat. no.:76266-138
Spécifications
A propos de cet article
The Optimus rack is compatible with CleanPak™ and TipEject™ reloads for easy transfer and storage.
- Double ball hinge allows the lid to open 180° for optimal multichannel pipetting
- Ribs in the base & snapcard interact to reduce snapcard flex for consistent liquid levels with multichannel devices
- Withstands repeatable autoclaving and reuse
- Locking clasp secures tips during transport and non-use
- Great stackability for easy storage
The Optimus rack is compatible with both uTIP™ and xTIP4™ pipette tip lines. The empty racks are available in two sizes; for tip volumes from 10 to 300 µl or 1000 to 1250 µl. Both racks are packaged 10 per pack, 4 packs per case.
Customers Who Bought This Also Bought