ContactUsLinkComponent
XTIP4* Pipette Tip Tipeject reload 250ul
xTIP4™ TipEject™ Pipette Tip Reload, Rainin® LTS® Pipette Compatible, Biotix
# de catalogue 76266-130
Fournisseur:  Biotix
   Waste Reducing
XTIP4* Pipette Tip Tipeject reload 250ul
xTIP4™ TipEject™ Pipette Tip Reload, Rainin® LTS® Pipette Compatible, Biotix
# de catalogue 76266-130
Fournisseur:  Biotix
   Waste Reducing
Spécifications
  • Volume:
    250 µl
  • Environmentally Preferable:
    Waste Reducing
  • Packed:
    Reload: 96 Tips/Refill; 10 Refills/Pack; 5 Packs/Case
  • Sterile:
    +
  • Tip Style:
    TipEject™ Reload
Spécifications
A propos de cet article
Create a perfect ergonomic system in the lab with FlexFit®, StarStop®, and a Biotix xPIPETTE® or Rainin LTS® pipette.
  • Clear casing and colored snap cards for easy tip identification
  • Operated easily with one hand, without directly contacting the tips
  • Reuse empty racks to reduce waste
  • Tips are shielded from the environment and user handling
  • Complete multi-channel and single-channel xPIPETTE® and LTS® pipette compatibility
Light Insertion Force
FlexFit® sealing technology requires minimal insertion force to generate consistent seals that users can trust with every tip.
Light Ejection
No matter how tips are applied, xTIP4™ pipette tips will always eject at the same light force. The StarStop® feature works with LTS-style pipettes to create a strong seal in the same location, every time.
Low Binding
Biotix low retention technology reduces the amount of sample lost in the tip when pipetting. Great for use with loading dyes, proteins, and DNA to increase an experiment's accuracy and yield.
Clean Delivery
These tips are engineered with the Blade® ratio to reduce hanging droplet formation when dispensing a sample. This limits the need for tip touch off on plate wells or tubes, saving time and reagent.
Certifications: ISO 9001:2015, Certified free of Dnase, RNnase, Endotoxins/pyrogens, and PCR inhibitors. Sterilized product is certified to 10⁻³ SAL.
BodyTextAccessories: Compatible Empty rack part numbers: 76266-138 and 76266-140.
Customers Who Bought This Also Bought