Spécifications
- Volume:1000 µl
- Environmentally Preferable:Waste Reducing
- Packed:Reload: 96 Tips/Refill; 8 Refills/Pack; 5 Packs/Case
- Sterile:+
- Tip Style:CleanPak™ Reload
Spécifications
A propos de cet article
Create a perfect ergonomic system in the lab with FlexFit®, StarStop®, and a Biotix xPIPETTE or Rainin LTS® pipette.
- Low cost option to get high-quality tips
- 960 tips on the lab bench using the same surface area as 1 rack
- Re-sealable bag is available in both sterile and nonsterile formats
- Color-coded snap cards allow for quick identification
- The product is fully recyclable and contains no contaminating cardboard or paper
- Inserts are easy to transfer to an Optimus empty rack
- Full multi-channel and single-channel xPIPETTE and LTS® pipette compatibility
Light Insertion Force
FlexFit® sealing technology requires minimal insertion force to generate consistent seals that users can trust with every tip.
Light Ejection
No matter how tips are applied, xTIP4™ pipette tips will always eject at the same light force. The StarStop® feature works with LTS-style pipettes to create a strong seal in the same location, every time.
Low Binding
Biotix low retention technology reduces the amount of sample lost in the tip when pipetting. Great for use with loading dyes, proteins, and DNA to increase an experiment's accuracy and yield.
Clean Delivery
These tips are engineered with the Blade® ratio to reduce hanging droplet formation when dispensing a sample. This limits the need for tip touch off on plate wells or tubes, saving time and reagent.