ContactUsLinkComponent
xTIP4™ Racked Pipette Tips, Rainin® LTS® Pipette Compatible, Biotix
Catalogue # 76266-106
Fournisseur:  Biotix
   Waste Reducing
   Sustainable Packaging
xTIP4™ Racked Pipette Tips, Rainin® LTS® Pipette Compatible, Biotix
Catalogue # 76266-106
Fournisseur:  Biotix
Supplier Number:  63300024
   Waste Reducing
   Sustainable Packaging
Spécifications
  • Volume:
    1200 µl
  • Environmentally Preferable:
    Waste Reducing,
    Sustainable Packaging
  • Environmental certification:
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • Packed:
    Racked: 768/Pack, 3840/Case
  • Cat. no.:
    76266-106
Spécifications
À propos de cet article
Biotix xTIP4 pipette tips are fully compatible with Rainin LTS single, multi, and electronic pipettes. Create a perfect ergonomic system in your lab with FlexFit®, StarStop®, and a Biotix xPIPETTE or Rainin LTS® pipette.
  • Polyethylene filters protect against aerosol and liquid contamination with no colorant or CMC contamination
  • Full multi-channel and single-channel LTS and xPIPETTE compatibility
Light Insertion Force
FlexFit® sealing technology requires minimal insertion force to generate consistent seals that users can trust with every tip.
Light Ejection
No matter how tips are applied, xTIP4™ pipette tips will always eject at the same light force. The StarStop® feature works with LTS-style pipettes to create a strong seal in the same location, every time.
Low Binding
Biotix low retention technology reduces the amount of sample lost in the tip when pipetting. Great for use with costly reagenst like proteins, and DNA to increase an experiment's accuracy and yield.
Clean Delivery
These tips are engineered with a Blade® tip to reduce hanging droplet formation when dispensing a sample. This limits the need for tip touch off on plate wells or tubes, saving time and sample.
Frequently Bought Together