Spécifications
- Description:UA Liquid Controls Plus
- Size:2×5 ml
- Cat. no.:76266-172
Spécifications
A propos de cet article
UA liquid controls plus is an assayed urine control intended for use in validating the precision of visual and analyzer reading of urinalysis.
Use for one or more of the following analytes: Ascorbic Acid, Glucose, Bilirubin, Ketone (Acetoacetic Acid), Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes, and Human Chorionic Gonadotropin.