ContactUsLinkComponent
Test Tube Rack 25mm / 24 tubes
Wet/Dry Plastic Test Tube Racks, United Scientific Supplies
# de catalogue 76263-384
Test Tube Rack 25mm / 24 tubes
Wet/Dry Plastic Test Tube Racks, United Scientific Supplies
# de catalogue 76263-384
Spécifications
  • Description:
    Wet/Dry Plastic Test Tube Rack
  • No. of Holes:
    21
  • Cat. no.:
    76263-384
  • For Tube O.D.:
    25 mm
Spécifications
A propos de cet article
Economical yet very durable rack.
  • Autoclavable
  • Easy to stack
These racks are submersible, autoclavable, and easily stack when empty. Racks are made from a special blend of polypropylene that makes them sink in a water bath and maintain their stability, even in agitated water. The top tier features an alphanumeric grid reference.
Packaging: Comes in packages of 6.
Customers Who Bought This Also Bought