Stainless Steel Micro Spoon, United Scientific Supplies
# de catalogue 76263-362
Fournisseur: United Scientific Supplies
Spécifications
- Description:Stainless Steel Micro Spoon
- Cat. no.:76263-362
Spécifications
A propos de cet article
Stainless steel micro spoon has an overall length of 23.5 cm (9.25"), with one end spoon and one end flat.
- Stainless steel design
- Micro spoon and flat edge
This micro spoon is perfect for mixing and dispensing small volume of powders or solutions.
Customers Who Bought This Also Bought