Slide Storage Boxes, United Scientific Supplies
# de catalogue 76263-278
Fournisseur: United Scientific Supplies
Spécifications
- Dimensions:230×184×32 mm
- Description:100 Place Slide Storage Box
- Cat. no.:76263-278
Spécifications
A propos de cet article
Store slides easily and securely.
- Engraved location numbers
- Polystyrene
Sturdy, two-piece molded polystyrene slide boxes hold standard 75×25 mm microscope slides. The bottom section has numbers engraved for easy reference, and the inside of the lid carries a numbered slide index, plastic.
Customers Who Bought This Also Bought