Wet/Dry Plastic Test Tube Racks, United Scientific Supplies
Catalogue # 76263-276
Fournisseur: United Scientific Supplies
Spécifications
- No. of Holes:40
- Description:Wet/Dry Plastic Test Tube Rack
- Cat. no.:76263-276
- For Tube O.D.:20 mm
Spécifications
À propos de cet article
Economical yet very durable rack.
- Autoclavable
- Easy to stack
These racks are submersible, autoclavable, and easily stack when empty. Racks are made from a special blend of polypropylene that makes them sink in a water bath and maintain their stability, even in agitated water. The top tier features an alphanumeric grid reference.
Packaging: Comes in packages of 6.
Frequently Bought Together