Utility Funnels, United Scientific Supplies
# de catalogue 76263-228
Fournisseur: United Scientific Supplies
Spécifications
- Capacity:—
- Description:Set of Three (1 of each Size)
- Height:—
- Material:PP
- Top I.D.:—
- Stem Length:—
- Stem Diameter:—
Spécifications
A propos de cet article
Useful for adding powder chemical to reaction vessels.
- Polypropylene
- Autoclavable
These funnels have eight internal ribs to facilitate filtration. They are made of polypropylene.
Customers Who Bought This Also Bought