Order Entry
ContactUsLinkComponent
RACK 5 PIPETTE SURESTAND 5MM ACRYLIC
SureStand™ Multi-Channel Capable Pipette Racks
# de catalogue 470303-334
Fournisseur:  MTC BIO, INC.
RACK 5 PIPETTE SURESTAND 5MM ACRYLIC
SureStand™ Multi-Channel Capable Pipette Racks
# de catalogue 470303-334
Fournisseur:  MTC BIO, INC.

Spécifications

  • Dimensions:
    10.5×9.5×5"
  • Description:
    Pipette Rack for 5 Pipettes (2 Multi-Channel)
  • Cat. no.:
    470303-334

Spécifications

A propos de cet article

SureStand™ Pipette racks accommodate multi-channel pipettes with the stability of a traditional acrylic rack. The “windows” allow the user to insert the multi-channel manifold through the rack, while keeping the body of the pipette stable by securing the pipette at two points; under the finger hook, and near the bottom of the handle.

  • Universal design; fits all popular pipettes
  • 5 mm acrylic construction is 40% thicker
  • Non-skid rubber feet for extra stability
  • Multi-channel 'window' slots

SureStand™ racks are available in 3 versions; a 3-place rack that accepts one multi-channel, a 5-place rack that accepts two multi-channels, and a 6-place rack that accepts four multichannels.

Each SureStand™ rack is also outfitted with four non-skid rubber feet for extra stability. SureStand™ will be the sturdiest and most versatile pipette rack in your lab.

Customers Who Bought This Also Bought